Description
- Easily tackle heavy cleaning tasks - this multipurpose cleaning cloth is both strong and absorbent.
- Cut cleaning time by up to 35% and use up to 31% less effort than rental towels, as the cloth's suitable for repeated use.
- Protect hands from heat and metal scraps while you clean, thanks to this cleaning cloth's heavy-duty fabric.
- Cut down on your use of solvents by up to 41% and waste by up to 85% versus rags with the exelCLEAN™ material.
Specifications
Specifications
| Product Type | Heavy-duty Wiper |
| Length (English) | 16.34 in. |
| Length (Metric) | 41.5 cm |
| Width (English) | 13.98 in. |
| Width (Metric) | 35.5 cm |
| Color | White |
| Ply | 1-ply |