Learn More
HEG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564935
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: NQEDFSTVSSK EGVMVQTSGK SHAASDAPEN LTLLAETADA RGRSGSSSRT NFTILPVGYS LEIATALTSQ SGNLASESLH LPSSSSEFDE RIAAFQTKSG TASEMGTERA MGLSEEWTV Highest antigen sequence identity to the following orthologs - mouse 43%, rat 38%.
Receptor component of the CCM signaling pathway which is a crucial regulator of heart and vessel formation and integrity May act through the stabilization of endothelial cell junctions.Specifications
HEG1 | |
Polyclonal | |
Unconjugated | |
HEG1 | |
KIAA1237 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57493 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULI3 | |
HEG1 | |
Recombit Human HEG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only