Learn More
HEG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552842
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NQEDFSTVSS KEGVMVQTSG KSHAASDAPE NLTLLAETAD ARGRSGSSSR TNFTILPVGY SLEIATALTS QSGNLASESL HLPSSSSEFD ERIAAFQTKS GTASEMGTER AMGLSEEWTV Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 38%.
Receptor component of the CCM signaling pathway which is a crucial regulator of heart and vessel formation and integrity May act through the stabilization of endothelial cell junctions. [UniProt]Specifications
HEG1 | |
Polyclonal | |
Unconjugated | |
HEG1 | |
4632417D23Rik; 5530401I02Rik; 9530025L16Rik; BC038614; Gm629; heart development protein with EGF like domains 1; heart development protein with EGF-like domains 1; heart of glass; heart of glass soluble isoform 1; heart of glass soluble isoform 2; heart of glass transmembrane isoform; HEG; HEG homolog 1; HEG homolog 1 (zebrafish); Heg_SE1; Heg_SE2; Heg_TM; HEG1; KIAA1237; MST112; MSTP112; OTTMUSG00000027315; protein HEG; protein HEG homolog 1; RGD1564641 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57493 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULI3 | |
HEG1 | |
Recombinant protein corresponding to Human HEG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only