Learn More
Heidolph™ Hei-VAP Core Allround Chill Package
Product Code. 16662284 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
EU Plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
16662284 EU Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 16662284 Supplier Heidolph™ Supplier No. 5712130000
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Hei-VAP Core with hand lift and vertical glassware set G3.
  • Hei-VAP Core with hand lift and vertical glassware set G3
  • Hei-VAC Control
  • Vacuum pump Rotavac Valve Tec
  • Tubing set
  • Chiller Hei-CHILL 250
  • Thermal fluid Kyro 30 (10 L)
TRUSTED_SUSTAINABILITY
Specifications
Type Hand Lift Rotary Evaporator
Plug Type EU Plug