Learn More
Heidolph™ Hei-VAP Reaction Extraction-Chill Package, EU-plug
Product Code. 17373330 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
EU-plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
17373330 EU-plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17373330 Supplier Heidolph™ Supplier No. 5742360000
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The Hei-VAP Reaction Extraction-Chill Package features a motor lift, chiller, and vapor temperature sensor.
  • Hei-VAP Expert Control with motor lift and vertical glassware set G6
  • Vacuum pump Rotavac Vario Tec
  • Vapor temperature sensor
  • Chiller Hei-CHILL 350
  • RS 232 cable
  • Thermal fluid Kyro 30 (10 L)
  • Tubing set
TRUSTED_SUSTAINABILITY
Specifications
Type Motor Lift Rotary Evaporator
Plug Type EU-plug