Learn More
Heidolph™ Hei-VAP University Research-Chill Package, EU-plug
Product Code. 17323380 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
EU-plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
17323380 EU-plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17323380 Supplier Heidolph™ Supplier No. 5762730500
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The Hei-VAP University Research-Chill Package features a motor lift, chiller, and tubing set.
  • Hei-VAP Ultimate Control with motor lift and vertical glassware set G3 XL
  • Vacuum pump Rotavac Vario Control
  • Chiller Hei-CHILL 600
  • RS 232 cable
  • Tubing set
  • Thermal fluid Kyro 30 (10 L)
TRUSTED_SUSTAINABILITY
Specifications
Type Motor Lift Rotary Evaporator
Plug Type EU-plug