Learn More
Heidolph™ Hei-VAP University Research Package, EU-plug
Product Code. 17259039 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
EU-plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
17259039 EU-plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17259039 Supplier Heidolph™ Supplier No. 5762630500
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The Hei-VAP University Research Package features a motor lift, and tubing set.
  • Hei-VAP Ultimate Control with motor lift and vertical glassware set G3 XL
  • Vacuum pump Rotavac Vario Control
  • Tubing set
TRUSTED_SUSTAINABILITY
Specifications
Type Motor Lift Rotary Evaporator
Plug Type EU-plug