Learn More
Macherey-Nagel™ Height Adjustable Manual Crimper
Product Code. 12738372 Shop All Macherey-Nagel Products
Change view
Click to view available options
Size:
20 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12738372 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12738372 Supplier Macherey-Nagel™ Supplier No. 735132
  • ONLINE EXCLUSIVE PRICE
    363.00€
    343.35€ / Pack of 1
    Save 19.65€
    5% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
No. per Pack 1 Each
Type Manual Crimper
For Use With (Equipment) For 20mm Flip Top/Flip Off Caps
Size 20 mm