missing translation for 'onlineSavingsMsg'
Learn More

Hematin, TRC

Product Code. 30138437
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138437 500 mg 500mg
30011605 1 g 1g
30003985 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138437 Supplier Toronto Research Chemicals Supplier No. TRCH235303500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15489-90-4
Molecular Formula C34H33N4O5Fe
Molecular Weight (g/mol) 633.49
Synonym Haematin, Hydroxy[dihydrogen Protoporphyrin IX-ato(2-)]iron, Hydroxyhemin, Phenodin, Protohematin, [Dihydrogen 3,7,12,17-tetramethyl-8,13-divinyl-2,18-porphinedipropionato(2-)]hydroxyiron, Ferrihemate, Ferriheme Hydroxide, Ferriporphyrin Hydroxide, Ferriprotoporphyrin IX Hydroxide, Ferriprotoporphyrin Basic, Growthphyllin
SMILES CC1=C(CCC(=O)O)C2=CC3=C(CCC(=O)O)C(=C4C=C5C(=C(C=C)C6=N5[Fe+3]7([OH-])(N2=C1C=C8C(=C(C)C(=C6)[N-]78)C=C)[N-]34)C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C34H34N4O4.Fe.H2O/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;;/h7-8,13-16H,1-2,9-12H2,3-6H3,(H4,35,36,37,38,39,40,41,42);;1H2/q;+3;/p-3/b25-13?,26-13-,27-14?,28-15-,29-14+,30-15?,31-16-,32-16?;;
Formula Weight 633.18
Chemical Name or Material Hematin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.