missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEMGN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17616-1-AP-20UL
This item is not returnable.
View return policy
Description
HEMGN regulates the proliferation and differentiation of hematopoietic cells. Overexpression of HEMGN block the TPA-induced megakaryocytic differentiation in the K562 cell model. HEMGN may also prevent cell apoptosis through the activation of the nuclear factor-kappa B (NF-kB).Specifications
| HEMGN | |
| Polyclonal | |
| Unconjugated | |
| HEMGN | |
| EDAG, EDAG 1, HEMGN, hemogen, Hemopoietic gene protein, NDR, PRO1620 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55363, 93966 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BXL5, Q9ERZ0 | |
| HEMGN | |
| HEMGN Fusion Protein Ag11793 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction