missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin A2 Antibody (rHBA/9167), Novus Biologicals™
Mouse Monoclonal Antibody
232.00€ - 462.00€
Specifications
| Antigen | Hemoglobin A2 |
|---|---|
| Clone | rHBA/9167 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18691795
|
Novus Biologicals
NBP3-23944-100ug |
100 μg |
462.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602265
|
Novus Biologicals
NBP3-23944-20ug |
20 μg |
232.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hemoglobin A2 Monoclonal antibody specifically detects Hemoglobin A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Hemoglobin A2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cardiovascular Biology | |
| 10 mM PBS with 0.05% BSA | |
| 3040 | |
| IgG2b κ | |
| Protein A or G purified |
| rHBA/9167 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| alpha globin, alpha-2 globin, Alpha-globin, Hemoglobin alpha chain, hemoglobin subunit alpha, hemoglobin, alpha 2 | |
| Recombinant fragment (around aa1-100) of human Hemoglobin A2 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title