missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin Antibody (2A1), Alexa Fluor™ 750 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20120AF750
This item is not returnable.
View return policy
Description
Hemoglobin Monoclonal antibody specifically detects Hemoglobin in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| Hemoglobin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| 2A1 | |
| 50mM Sodium Borate | |
| Murine hybridomas were generated using spleen cells from aging (8-10 month old) NZB/W F1 animals. Hybridomas secreting antibodies reactive to murine hemoglobin were subcloned by limiting dilution and antibody isotypes determined using commercial reagents. Eight-week-old NZB/W F1 were immunized subcutaneously with 50 mg mHb antigen emulsified in Complete Fruend's Adjuvant. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction