missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ HepExtend™ Media Kit
Description
The HepExtend™ Media Kit contains the new HepExtend Supplement (50X), as well as William's E Medium and Primary Hepatocyte Maintenance Supplement, providing the three components needed to optimize long term hepatocyte culture. HepExtend Supplement has been optimized for use with cryopreserved primary hepatocytes to improve cell viability, function and number of days in culture. This enables researchers to perform metabolic and toxicity experiments not achievable using standard culture conditions. Simply add the HepExtend Supplement and Primary Hepatocyte Maintenance Supplement to the William’s E Medium for a complete solution. The HepExtend supplement does not contain any small molecules or fetal bovine sera, components that are known to interfere with primary hepatocyte function.
Enabling—offers maximum cell viability and cell health when culturing primary plateable hepatocytes
Extended cell survival—days in culture can be extended to >10 days while maintaining normal cell morphology and bile canaliculi
Consistent—manufactured in a cGMP compliant facility using high quality materials
Maintain P450 activity
The HepExtend Media Kit can be used to help ensure metabolically competent hepatocytes for at least 10 days for use in downstream ADME/Tox applications not achieved with standard 5-day culture systems.
Enhanced gene expression
Using the components in the HepExtend Media Kit to culture primary human hepatocytes increases the basal gene expression of many key genes involved in drug metabolism and transport.
Retain inducibility
Nuclear receptor activation and gene inducibility are retained in cultures treated with HepExtend supplemented media.
Maintain P450 activity
The HepExtend Media Kit can be used to help ensure metabolically competent hepatocytes for at least 10 days for use in downstream ADME/Tox applications not achieved with standard 5-day culture systems.
Enhanced gene expression
Using the components in the HepExtend Media Kit to culture primary human hepatocytes increases the basal gene expression of many key genes involved in drug metabolism and transport.
Retain inducibility
Nuclear receptor activation and gene inducibility are retained in cultures treated with HepExtend supplemented media.
Specifications
Specifications
| Cell Type | Mammalian Cells |
| Form | Cryopreserved |
| Product Type | Cell Culture Medium |
| Product Line | HepExtend |
| Quantity | 1 Kit |
| Shipping Condition | Dry Ice |
| Content And Storage | • 1 William's E Medium (A1217601) • 1 Primary Hepatocyte Maintenance Supplements (CM4000) • 1 HepExtend™ Supplement (50X) (A2737501) Store medium at 2–8°C degrees and supplements at -5 to -30°C. |
| Culture Type | Mammalian Cell Culture |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?