missing translation for 'onlineSavingsMsg'
Learn More

Melford HEPPSO

Product Code. 30018346
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 30018346

Brand: Melford H7507025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HEPPSO (N-(2-Hydroxyethyl)piperazine-N'-(3-propanesulfonic acid)) is a zwitterionic buffering agent commonly used particularly in biochemical and molecular biology research.

HEPPSO (N-(2-Hydroxyethyl)piperazine-N'-(3-propanesulfonic acid)) is a zwitterionic buffering agent commonly used particularly in biochemical and molecular biology research. It is valued for its ability to maintain a stable pH in a variety of biological assays and experiments.

HEPPSO is often preferred for its buffering capacity over a wide pH range, typically between pH 6.8 to 8.2, making it suitable for various applications such as protein purification, enzyme assays, cell culture, and electrophoresis.

Additionally, HEPPSO is less likely to interfere with biological processes compared to some other buffering agents, making it a versatile choice for maintaining optimal conditions in biological experiments.

Specifications

Chemical Name or Material HEPPSO
CAS 68399-78-0
Quantity 25 g
Molecular Formula C9H20N2O5S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.