Learn More
Heptachlor, TRC
Specifications
Specifications
| Chemical Name or Material | Heptachlor |
| CAS | 76-44-8 |
| Molecular Formula | C10 H5 Cl7 |
| Quantity | 50 mg |
| Synonym | 1,4,5,6,7,8,8-Heptachloro-3a,4,7,7a-tetrahydro-4,7-methano-1H-indene, 1,4,5,6,7,10,10-Heptachloro-4,7,8,9-tetrahydro-4,7-methyleneindene, 1,4,5,6,7,8,8-Heptachloro-3a,4,7,7a-tetrahydro-4,7-methanoindene, 1,4,5,6,7,8,8-Heptachloro-3α,4,7,7α-tetrahydro-4,7-endo-methanoindene, 3-Chlorochlordene, Aahepta, Agroceres, Arbinex 30TN, E 3314, ENT 15,152, GPKh, Hepta, Heptachlor, Heptachlorane, Heptox, NSC 8930, Rhodiachlor, Velsicol 104 |
| InChI Formula | InChI=1S/C10H5Cl7/c11-4-2-1-3-5(4)9(15)7(13)6(12)8(3,14)10(9,16)17/h1-5H/t3?,4?,5?,8-,9+/m0/s1 |
| SMILES | ClC1C=CC2C1[C@@]3(Cl)C(=C(Cl)[C@]2(Cl)C3(Cl)Cl)Cl |
| IUPAC Name | 1,4,5,6,7,8,8-Heptachloro-3a,4,7,7a-tetrahydro-4,7-methano-1H-indene |
| Molecular Weight (g/mol) | 373.32 |
| Formula Weight | 369.8211 |