missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanoic Acid, TRC

Product Code. 17969634
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17969634 100 g 100g
17999074 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17969634 Supplier Toronto Research Chemicals Supplier No. TRCH281030100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-12-7
Molecular Formula C17 H34 O2
Molecular Weight (g/mol) 270.45
Synonym Heptadecanoic acid (8CI, 9CI, ACI), Margaric acid, Margarinic acid, n-Heptadecanoic acid, n-Heptadecoic acid, n-Heptadecylic acid, NSC 3743
IUPAC Name heptadecanoic acid
SMILES CCCCCCCCCCCCCCCCC(=O)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C17H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19/h2-16H2,1H3,(H,18,19)
Formula Weight 270.2559
Chemical Name or Material Margaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.