Learn More
Heptamethyltrisiloxane, TRC
Product Code. 30011127
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011127 10 mL 10mL
30010286 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30011127 Supplier Toronto Research Chemicals Supplier No. TRCH28113010ML
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1873-88-7
Molecular Formula C7 H22 O2 Si3
Molecular Weight (g/mol) 222.5
Synonym 1,1,1,3,5,5,5-Heptamethyltrisiloxane (ACI), 1,1,1,3,5,5,5-Heptamethylsiloxane, 2,2,4,6,6-Pentamethyl-3,5-dioxa-2,4,6-trisilaheptane, 2-Hydro-1,1,1,2,3,3,3-heptamethyltrisiloxane, Bis(trimethylsiloxy)methylsilane, Bis(trimethylsilyloxy)methylsilane, Heptamethylhydrotrisiloxane, Methylbis(trimethylsiloxy)silane, Methylbis(trimethylsilyloxy)silane, SIB 1844, SIB 1844.0
IUPAC Name trimethyl-[methyl(trimethylsilyloxy)silyl]oxysilane
Specifications
Quantity 10 mL
InChI Formula InChI=1S/C7H22O2Si3/c1-10(8-11(2,3)4)9-12(5,6)7/h10H,1-7H3
SMILES (2,5-dichlorophenyl)sulfanylmethylsulfanyl-diethoxy-sulfanylidene-λ^{5}-phosphane
Formula Weight 222.0928
Chemical Name or Material Heptamethyl Trisiloxane 1,1,1,3,5,5,5-Heptamethyltrisiloxane