missing translation for 'onlineSavingsMsg'
Learn More

HER2 Trastuzumab297.rMAb anti-Human, BV605, Clone: Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb), BD OptiBuild™

Product Code. 32228000
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32228000 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32228000 Supplier BD Biosciences Supplier No. 756618
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Biosimilar Monoclonal Antibody

HER2 Trastuzumab297.rMAb Monoclonal antibody specifically detects HER2 Trastuzumab297.rMAb in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HER2 Trastuzumab297.rMAb
Applications Flow Cytometry
Classification Monoclonal
Clone Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb)
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD340; ERBB2; HER-2; HER-2/neu; HER2; NEU; proto-oncogene c-ErbB-2
Immunogen A431 Cell Line Expressing EGFR
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2064
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.