missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27459-1-AP-150UL
This item is not returnable.
View return policy
Description
HERC2 Polyclonal antibody apecifically detetects HERC2 antigen in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ELISA.Specifications
| HERC2 | |
| Polyclonal | |
| Unconjugated | |
| BC047911 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 250 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| HERC2 | |
| HERC2 fusion protein Ag26501 | |
| 150 μL | |
| 8924 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction