missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 2 Antibody (HSVA33), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-2515
This item is not returnable.
View return policy
Description
Herpes Simplex Virus 2 Monoclonal antibody specifically detects Herpes Simplex Virus 2 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Herpes Simplex Virus 2 | |
| Monoclonal | |
| Unconjugated | |
| Herpes Simplex Virus Type 2 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified HSV from strain BH. | |
| 0.2 mg | |
| Virology Bacteria and Parasites | |
| Reacts with 56-64 kD band corresponding to glycoprotein D band in immunoblotting of purified HSV under reduced conditions and with 120-140 kD band in immunoblotting of HSV-infected Vero cells lysates under non-reduced conditions. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| Western Blot, ELISA | |
| HSVA33 | |
| Western Blot 1:100-1:2000, ELISA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction