missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus UL42 Antibody (2H4), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2762AF532
This item is not returnable.
View return policy
Description
HSV UL42 Monoclonal specifically detects HSV UL42 in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| HSV UL42 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| Virology Bacteria and Parasites | |
| May recognize UL42 bound to Herpes Simplex Virus polymerase. Recognizes amino acids 360-366 in UL42. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 2H4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| HHV1gp061 | |
| Baculovirus-expressed HSV DNA polymerase (POL) and POL/UL42 complex. | |
| 0.1 mL | |
| Primary | |
| Non-species specific | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction