missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus UL42 Antibody (2H4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
214.00€ - 498.00€
Specifications
| Antigen | Herpes Simplex Virus UL42 |
|---|---|
| Clone | 2H4 |
| Concentration | 1.1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18576251
|
Novus Biologicals
NB100-2762-0.025mg |
0.025 mg |
214.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18550532
|
Novus Biologicals
NB100-2762 |
0.1 mg |
498.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Herpes Simplex Virus UL42 Monoclonal antibody specifically detects Herpes Simplex Virus UL42 in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| Herpes Simplex Virus UL42 | |
| 1.1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| HHV1gp061 | |
| IgG1 | |
| Protein A purified |
| 2H4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Virology Bacteria and Parasites | |
| PBS | |
| Baculovirus-expressed Herpes Simplex Virus DNA polymerase (POL) and POL/UL42 complex. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title