missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HES5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523498
This item is not returnable.
View return policy
Description
HES5 Polyclonal Antibody for Western Blot, IHC (P)
Hairy and Enhancer-of-split homologues 5 (HES5) isa helix-loop-helix transcriptional factor that negatively regulates cell differentiation during embryogenesis. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. HES5 is reported to be a down stream effector of the Notch Signaling Pathway and is expressed in advanced ovarian serous adenocarcinomas as well as in cervical squamous carcinoma. Reports also suggest that HES5 is abundantly expressed in osteoarthritis along with Notch1 and Jagged1.
Specifications
| HES5 | |
| Polyclonal | |
| Unconjugated | |
| HES5 | |
| BHLHB38; Class B basic helix-loop-helix protein 38; hairy and enhancer of split 5; hairy and enhancer of split 5 (Drosophila); hes family bHLH transcription factor 5; Hes5; Hes-5; RP3-395M20.5; Transcription factor HES-5 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 15208, 388585, 79225 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P70120, Q03062, Q5TA89 | |
| HES5 | |
| A portion of amino acid1-50 of human HES5. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction