missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Hexacosanoic acid, 97%

Product Code. 15427218
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427218 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15427218 Supplier Alfa Aesar™ Supplier No. H60726.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 506-46-7
Melting Point 87°C to 89°C
Molecular Formula C26H52O2
Linear Formula CH3(CH2)24COOH
MDL Number MFCD00002811
Quantity 250 mg
Beilstein 1799681
Synonym cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
InChI Key XMHIUKTWLZUKEX-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
IUPAC Name hexacosanoic acid
Molecular Weight (g/mol) 396.7
PubChem CID 10469
ChEBI CHEBI:31009
Formula Weight 396.69
Percent Purity 97%
Chemical Name or Material Hexacosanoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.