missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanoic-12-d1 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic-12-D Acid |
| CAS | 358730-99-1 |
| Molecular Formula | C16 D H31 O2 |
| Quantity | 0.1 g |
| Synonym | Hexadecanoic-12-d acid (9 CI), Hexadecanoic-12-D Acid, Palmitic acid-12-d, [12-2H1]Hexadecanoic acid, [12-2H1]Palmitic acid, Hexadecanoic Acid-12-d |
| InChI Formula | InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i5D |
| SMILES | [2 H]C(CCCC)CCCCCCCCCCC(=O)O |
| IUPAC Name | 12-deuteriohexadecanoic acid |
| Molecular Weight (g/mol) | 257.4302 |
| Formula Weight | 257.2465 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction