missing translation for 'onlineSavingsMsg'
Learn More

Hexafluoroglutaric anhydride, 97%, Thermo Scientific Chemicals

Product Code. 11357845
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357845 25 g 25g
11337845 1 g 1g
11347845 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357845 Supplier Thermo Scientific Alfa Aesar Supplier No. L02798.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-68-1
Molecular Formula C5F6O3
Molecular Weight (g/mol) 222.042
MDL Number MFCD00006680
InChI Key IHYAGCYJVNHXCT-UHFFFAOYSA-N
Synonym hexafluoroglutaric anhydride, perfluoroglutaric anhydride, 3,3,4,4,5,5-hexafluorodihydro-2h-pyran-2,6 3h-dione, hexafluoroglutaric acid anhydride, 2,2,3,3,4,4-hexafluoropentanedioic anhydride, 3,3,4,4,5,5-hexafluoro-3h-4,5-dihydropyran-2,6-dione, hexafluoroglutaricanhydride, acmc-1clz4, hexafluorooxane-2,6-dione
PubChem CID 78995
IUPAC Name 3,3,4,4,5,5-hexafluorooxane-2,6-dione
SMILES C1(=O)C(C(C(C(=O)O1)(F)F)(F)F)(F)F

Specifications

Density 1.652
Boiling Point 71°C to 72°C
Flash Point None
Odor Pungent
Refractive Index 1.324
Quantity 25 g
UN Number UN3265
Beilstein 210699
Sensitivity Moisture sensitive
Formula Weight 222.04
Percent Purity 97%
Chemical Name or Material Hexafluoroglutaric anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.