missing translation for 'onlineSavingsMsg'
Learn More

Hexakis(2,2-difluoroethoxy)phosphazene, TRC

Product Code. 30133998
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133998 50 mg 50mg
30132580 100 mg 100mg
30004816 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133998 Supplier Toronto Research Chemicals Supplier No. TRCH29402850MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186817-57-2
Molecular Formula C12H18F12N3O6P3
Molecular Weight (g/mol) 621.19
Synonym Hexakis(1H,1H,2H-perfluoroethoxy)phosphazene;2,2,4,4,6,6-Hexakis(2,2-difluoroethoxy)-2,2,4,4,6,6-hexahydro-1,3,5,2,4,6-triazatriphosphorine
IUPAC Name 2,2,4,4,6,6-hexakis(2,2-difluoroethoxy)-1,3,5-triaza-2λ^{5},4λ^{5},6λ^{5}-triphosphacyclohexa-1,3,5-triene
SMILES FC(F)COp1(OCC(F)F)np(OCC(F)F)(OCC(F)F)np(OCC(F)F)(OCC(F)F)n1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H18F12N3O6P3/c13-7(14)1-28-34(29-2-8(15)16)25-35(30-3-9(17)18,31-4-10(19)20)27-36(26-34,32-5-11(21)22)33-6-12(23)24/h7-12H,1-6H2
Formula Weight 621.0217
Percent Purity >95%
Chemical Name or Material Hexakis(2,2-difluoroethoxy)phosphazene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.