missing translation for 'onlineSavingsMsg'
Learn More

Hexanoic Anhydride, TRC

Product Code. 30397339
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397339 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397339 Supplier Toronto Research Chemicals Supplier No. TRCH29433550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexanoic Anhydride
CAS 2051-49-2
Molecular Formula C12H22O3
Quantity 50 g
Synonym 1,1'-Anhydride Hexanoic Acid, Hexanoic Acid Anhydride, Caproic Acid Anhydride, Caproic Anhydride, Capronic Acid Anhydride, Capronic Anhydride, Hexanoyl Anhydride, n-Hexanoic Acid Anhydride, n-Hexanoic Anhydride
InChI Formula InChI=1S/C12H22O3/c1-3-5-7-9-11(13)15-12(14)10-8-6-4-2/h3-10H2,1-2H3
SMILES CCCCCC(=O)OC(=O)CCCCC
IUPAC Name hexanoyl hexanoate
Molecular Weight (g/mol) 214.3
Formula Weight 214.1569
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.