missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Acetate, TRC

Product Code. 30406668
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406668 10 g -
30401545 100 g 100g
30400157 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406668 Supplier Toronto Research Chemicals Supplier No. TRCH29640010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-92-7
Molecular Formula C8 H16 O2
Molecular Weight (g/mol) 144.21
Synonym Hexyl alcohol, acetate (6CI), 1-Hexyl acetate, Exceed 600, Hexyl acetate, Hexyl ester acetic acid, Hexyl ethanoate, NSC 7323, ZED-N 60, n-Hexyl acetate, n-Hexyl ethanoate, Acetic acid, hexyl ester, Acetic Acid-hexyl Ester, Hexyl alcohol, acetate (6CI), 1-Hexyl acetate, Exceed 600, Hexyl acetate, Hexyl ester acetic acid, Hexyl ethanoate, NSC 7323, ZED-N 60, n-Hexyl acetate, n-Hexyl ethanoate, Acetic acid, hexyl ester, Acetic Acid-hexyl Ester
IUPAC Name hexyl acetate
SMILES CCCCCCOC(=O)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H16O2/c1-3-4-5-6-7-10-8(2)9/h3-7H2,1-2H3
Formula Weight 144.115
Chemical Name or Material Hexylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.