Learn More
LNI Swissgas HG 2U PRO 350
Product Code. 17852329 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
17852329 100/115/220/230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17852329 Supplier LNI Swissgas Supplier No. 6920.16.035
  • ONLINE EXCLUSIVE PRICE
    12290.00€
    11191.32€ / Pack of 1
    Save 1098.68€
    8% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Hydrogen generator ultra high purity.
  • Accessory
Specifications
Voltage 100/115/220/230 V
Type Hydrogen Carrier Gas Generator