Learn More
Invitrogen™ HHV-6 Capsid Monoclonal Antibody (F24H)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA17043
Product Code. 11594503
  • 520.00€ / 1mL
Description
MA1-7043 detects the human Herpes Virus, type 6. This antibody is specific for the 140kDa capsid polypeptide of HHV6. MA1-7043 has been successfully used in immunoprecipitation, immunofluorescence and Western blot procedures. The MA1-7043 immunogen is Human Herpes Virus type 6 lysate.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HHV-6 Capsid
Monoclonal
0.1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Virus
Antibody
IgG1
Immunohistochemistry, Immunoprecipitation
F24H
Unconjugated
HHV-6
Human Herpes Virus type 6 lysate.
1 mL
Primary
4°C