Learn More
FTM Technologies™ HI3P Portable Fume Hood
Product Code. 11788004
Change view
Click to view available options
Flow Rate:
50 to 450 m3/h
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
11788004 50 to 450 m3/h Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11788004 Supplier FTM Technologies™ Supplier No. HI3P
  • ONLINE EXCLUSIVE PRICE
    1350.00€
    1080.76€ / Pack of 1
    Save 269.24€
    19% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Offers superior protection and safety in a compact and portable unit. FTM Technologies™ HI3P Portable Fume Hoods have integrated lighting for visibility.
  • Applications include product handling, welding, application of solvents, and more
  • Can be used as a recirculating (ductless) fume hood (active carbon filtration or dust filter) or with an external output (external output filter and exhaust duct)
Specifications
Dimensions (L x W x H) 350 x 200 x 430mm
Flow Rate 50 to 450 m3/h
Voltage 230 V
Weight (Metric) 6 kg
Frequency 50 Hz