Learn More
FTM Technologies™ HI5VP Extraction Booths
Product Code. 11758054
Change view
Click to view available options
Flow Rate:
50 to 450 m3/h
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
11758054 50 to 450 m3/h Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11758054 Supplier FTM Technologies™ Supplier No. HI5VP
  • ONLINE EXCLUSIVE PRICE
    3465.00€
    3371.02€ / Pack of 1
    Save 93.98€
    2% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Used for filtration needs where a channelled space is needed. FTM Technologies™ HI5VP Extraction Booths have shaped, shatterproof glass measuring 6mm thick.
  • Front of booth entirely open
  • Applications include chemical handling, spraying, pulverization and varnishing
  • Also suitable for cleaning solvents, tubs, or machines producing a large amount of fumes
  • Can be used as recycling or ductless - filtration with active carbon or dust filter
  • Can also be used with external evacuation (filter for evacuation plus exhaust duct)
Specifications
Dimensions (L x W x H) 800 x 700 x 700 mm
Flow Rate 50 to 450 m3/h
Voltage 230 V
Weight (Metric) 33 kg
Frequency 50 Hz