Learn More
HICE1 Polyclonal Antibody
GREENER_CHOICE
HICE1 Polyclonal Antibody
Brand:  Invitrogen PA545046
Product Code. 15556782
  • 530.00€ / 100µL
Description
HICE1 Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HICE1
Polyclonal
Unconjugated
HAUS8
2410004L22Rik; 4HAUS augmin-like complex, subunit 8; DGT4; HAUS augmin like complex subunit 8; HAUS augmin-like complex subunit 8; HAUS augmin-like complex, subunit 8; HAUS8; HEC1/NDC80 interacting, centrosome associated 1; HEC1/NDC80-interacting centrosome-associated protein 1; Hec1-interacting and centrosome-associated 1; Hice1; MGC20533; NY SAR 48; NYSAR48; NY-SAR-48; Sarcoma antigen NY-SAR-48; sarcoma antigen NY-SAR-48 homolog
Rabbit
Affinity Chromatography
RUO
93323
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9BT25
HAUS8
Synthetic peptide directed towards the N-terminal of human HAUS8 (aa 57-106).