Learn More
Abeba™ High Original 5004 Shoes
ESD dissipative shoe includes PU sole with high slip resistance.
Brand:  Abeba™ 5004/42
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15671758
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white
  • Slip resistance PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
  • Narrow fit
  • Wedge heel 45 mm
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
5004
White
42
Polyurethane