Learn More
Abeba™ High Original 5009 Shoes
ESD dissipative shoe includes PU sole with high slip resistance.
Brand: Abeba™ 5009/40
Additional Details : Weight : 0.55000kg
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather, perforated
- Black
- Slip resistance PU outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
- Leather top layer
- Narrow fit
- Wedge heel 45mm
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Slip-resistant | |
5009 |
Black | |
40 | |
Polyurethane |