Learn More
Abeba™ High Original 7002 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand:  Abeba™ 7002/36
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15662448
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather top layer
  • Narrow fit
  • Wedge heel 45 mm
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
7002
White
36
Polyurethane
Animal fats, oil and fuel