Learn More
Idex High-Pressure, Zero Dead Volume Unions
Product Code. 13010410
Change view
Click to view available options
Diameter (English) Inner:
0.01 in.
0.02 in.
Swept Volume:
0.071 μL
0.283 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Swept Volume unitSize
13010410 0.01 in. 0.071 μL Each
13040390 0.02 in. 0.283 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13010410 Supplier Idex Supplier No. 0201437
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
PEEK ZDV unions provide accurate results
  • Inert PEEK fluid path
  • 10-32 coned configuration
Specifications
Type Union
Diameter (English) Inner 0.01 in.
Pressure Rating 6000 psi
Swept Volume 0.071 μL
Quantity Each
Material PEEK
For Use With (Equipment) 1/16 in. OD Tubing