Description
- Each lot of ddNTPs is functionally tested to meet or exceed criteria for high-quality sequencing with Thermo Sequenase™ DNA polymerase
- High purity: >98% triphosphate
- In a convenient set containing ddATP, ddCTP, ddGTP, and ddTTP in separate tubes
Dideoxy sequencing, Genotyping reactions
Specifications
Specifications
| Format | Liquid |
| Concentration | 5 mM |
| Purity | ≥98% |
| Quantity | 4 x 0.5 μm |
| Color | Colorless |
| For Use With (Application) | Dideoxy sequencing, genotyping reactions |