Learn More
Nabertherm™ High-Temperature Furnaces with Molybdenum Disilicide Heating Elements and Fiber Insulation
Product Code. 17397839 Shop All Nabertherm Products
Change view
Click to view available options
Capacity (Metric):
8 L
16 L
Temperature (Metric):
1600°C
1750°C
1800°C
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Temperature (Metric) Unit Size
17397839 16 L 1800°C Each
17347839 8 L 1600°C Each
17357839 8 L 1750°C Each
17367839 8 L 1800°C Each
17377839 16 L 1600°C Each
17387839 16 L 1750°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17397839 Supplier Nabertherm™ Supplier No. HT0358VNA
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Due to their solid construction and compact stand-alone design, these high-temperature furnaces are perfect for processes in the laboratory where the highest precision is needed. Oustanding temperature uniformity and practical details set very high quality benchmarks.
  • Dual shell housing with fan cooling for low shell temperatures
  • Heating from both sides via molybdenum disilicide heating elements
  • High-quality fiber insulation backed by special insulation
  • Side insulation constructed with tongue and groove blocks provides for low heat loss to the outside
  • Long-life roof insulation with special suspension
  • Chain-guided parallel swivel door for defined opening and closing of the door
  • Vapor vent in the furnace roof with motorized exhaust air flaps, controlled via the extra function of the controller
  • Heating elements switched via thyristors
Please Note: A furnace below 2.9 kW will be delivered with a standard UK plug. Anything above 2.9 kW will be delivered with the appropriate CEE plug. For anything above 2.9 kW, please ensure you have the required socket prior to purchase.
Specifications
Controller Type P570
Capacity (Metric) 16 L
Temperature (Metric) 1800°C
Includes Molybdenum Disilicide Heating Elements and Fiber Insulation
Dimensions (D x W x H) Interior 300 x 200 x 260 mm
Dimensions (L x W x H) Exterior 690 x 820 x 1860 mm
Voltage 400 V
Wattage 12500 W