Learn More
HINT1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1G3E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67583-100UL
Description
The HINT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HINT1. This antibody recognizes human antigen. The HINT1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| HINT1 | |
| 1G3E4 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2a |
| Immunofluorescence | |
| 1000 μg/mL | |
| FLJ30414, FLJ32340, HINT, HINT1, PKCI 1, PKCI1, PRKCNH1, Protein kinase C inhibitor 1 | |
| 100 μL | |
| 3094 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |