missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hip Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00064399-D01P
This item is not returnable.
View return policy
Description
Hip Polyclonal antibody specifically detects Hip in Human samples. It is validated for Western Blot
Specifications
| Hip | |
| Polyclonal | |
| PBS (pH 7.4) | |
| FLJ90230, hedgehog interacting protein, hedgehog-interacting protein, HIPFLJ20992 | |
| HHIP (NP_071920.1, 1 a.a. - 700 a.a.) full-length human protein. MLKMLSFKLLLLAVALGFFEGDAKFGERNEGSGARRRRCLNGNPPKRLKRRDRRMMSQLELLSGGEMLCGGFYPRLSCCLRSDSPGLGRLENKIFSVTNNTECGKLLEEIKCALCSPHSQSLFHSPEREVLERDLVLPLLCKDYCKEFFYTCRGHIPGFLQTTADEFCFYYARKDGGLCFPDFPRKQVRGPASNYLDQMEEYDKVEEISRKHKHNCFCIQEVVSGLRQPVGALHSGDGSQRLFILEKEGYVKILTPEGEIFKEPYLDIHKLVQSGIKGGDERGLLSLAFHPNYKKNGKLYVSYTTNQERWAIGPHDHILRVVEYTVSRKNPHQVDLRTARVFLEVAELHRKHLGGQLLFGPDGFLYIILGDGMITLDDMEEMDGLSDFTGSVLRLDVDTDMCNVPYSIPRSNPHFNSTNQPPEVFAHGLHDPGRCAVDRHPTDININLTILCSDSNGKNRSSARILQIIKGKDYESEPSLLEFKPFSNGPLVGGFVYRGCQSERLYGSYVFGDRNGNFLTLQQSPVTKQWQEKPLCLGTSGSCRGYFSGHILGFGEDELGEVYILSSSKSMTQTHNGKLYKIVDPKRPLMPEECRATVQPAQTLTSECSRLCRNGYCTPTGKCCCSPGWEGDFCRTAKCEPACRHGGVCVRPNKCLCKKGYLGPQCEQVDRNIRRVTRAGILDQIIDMTSYLLDLTSYIV | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| NP_071920.1 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 64399 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction