Learn More
Cytiva His Mag Sepharose™ Ni Magnetic Beads
Product Code. 11545804 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1 mL
100 mL
10 x 1 mL
Unit Size:
100mL
10mL
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11545804 10 x 1 mL 10mL
11540894 5 x 1 mL Pack of 5
17296876 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11545804 Supplier Cytiva Supplier No. 28979917
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Designed for efficient, small-scale purification/screening of histidine-tagged proteins from different sources Cytiva Lifescience™ His Mag Sepharose™ Ni Magnetic Beads enable high-capacity small-scale purification.
Together with MagRack 6, a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel. For higher throughput, larger numbers of samples can easily be screened in parallel on a robotic device.
  • High-capacity small-scale purification of histidine-tagged proteins from different sources
  • High purity and yield
  • Easy parallel screening of histidine-tagged proteins with high repeatability
  • Scalability dash (en) Simple capture of histidine-tagged proteins from small or large sample volumes (low microliter to milliliter scale)
Order Info
1mL of medium slurry is sufficient for five purification runs according to the recommended protocol
TRUSTED_SUSTAINABILITY
Specifications
Binding Property 50 mg Histidine-tagged protein/mL medium
Quantity 10 x 1 mL
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3