missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen IXSelect

Product Code. 15569261 Shop All Cytiva Products
Change view
Click to view available options
Packaging Quantity:
4.7 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity unitSize
15569261 4.7 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15569261 Supplier Cytiva Supplier No. 17371410
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

HiScreen IXSelect columns are prepacked with IXSelect affinity chromatography medium, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.

  • IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography resin using the same linear fluid velocity. 
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 x 4.7 mL
For Use With (Application) Group specific affinity
Height (Metric) Bed 10 cm
Packaging Quantity 4.7 mL
compliance-icons
Nombre del producto
  • HiScreen IXselect
Palabra de advertencia
  • Warning
Clasificación
  • Flammable liquid Category 3
Indicaciones de peligro
  • H226-Flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Otros peligros
  • MIXTURE LIST-Contain: ethanol
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.