missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2BC Antibody (CL5048) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61153MFV500
This item is not returnable.
View return policy
Description
HIST1H2BC Monoclonal antibody specifically detects HIST1H2BC in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| HIST1H2BC | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| H2B histone family, member K, H2B/a, H2B/g, H2B/kH2B/h, H2BFA, H2BFG, H2BFH, H2BFKH2B/l, H2BFL, HIST1H2BC, HIST1H2BE, HIST1H2BF, HIST1H2BG, histone 1, H2bi, histone cluster 1, H2bi, histone H2B type 1-C/E/F/G/I, Histone H2B.1 A, Histone H2B.a, Histone H2B.g, Histone H2B.h, Histone H2B.k, Histone H2B.l | |
| This antibody was developed using a synthetic peptide derived from P62807, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL5048 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 8347 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction