missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
220.00€ - 789.00€
Specifications
| Conjugate | Unconjugated |
|---|---|
| Gene Alias | 2 (4 imidazolyl)ethylamine, 4 (2 aminoethyl) 1 3 diazole |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18776743
|
Novus Biologicals
NBP2-45266-0.1ml |
538.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18087933
|
Novus Biologicals
NBP2-45266-0.2ML |
0.2 mL |
789.00€
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667265
|
Novus Biologicals
NBP2-45266-0.025ml |
0.025 mL |
220.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifically detects Histamine in All samples, and it is validated for Dot Blot, Immunohistochemistry (Paraffin)
Histamine Polyclonal specifically detects Histamine in Human, All Species samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot.Specifications
| Unconjugated | |
| 2 (4 imidazolyl)ethylamine, 4 (2 aminoethyl) 1 3 diazole |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title