missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histone 1F0 Recombinant Rabbit Monoclonal Antibody (1C7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560618
This item is not returnable.
View return policy
Description
Histone 1F0 Recombinant Monoclonal Antibody for ELISA
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a replication-independent histone that is a member of the histone H1 family.
Specifications
| Histone 1F0 | |
| Recombinant Monoclonal | |
| 0.333 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P07305 | |
| H1-0, H1F0 | |
| A synthesized peptide. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 1C7 | |
| Unconjugated | |
| H1-0 | |
| D130017D06Rik; H1 histone family member 0; H1 histone family, member 0; H1(0); H1.0, H1(0), H1-0; H10; H1-0 histone; H1f0; H1fv; histone H1'; Histone H1(0); histone H1.0; Histone H1.0, N-terminally processed; Histone H1-0; MyD196 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3005 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction