missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-56346
This item is not returnable.
View return policy
Description
Histone H2a Polyclonal specifically detects Histone H2a in Human, Mouse, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Histone H2a | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| H2A histone family, member Q, H2A/q, H2AFQH2A, H2A-GL101, histone 2, H2ac, histone cluster 2, H2ac, histone H2A type 2-C, Histone H2A/q, Histone H2A-GL101, histone IIa, MGC74460 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| A 14 kDa band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 32849530), ELISA 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml | |
| P04908 | |
| HIST2H2AC | |
| This antibody was generated by immunizing rabbits with a mixture of synthetic peptides containing amino acid residues 1-15 (MSGRGKQGGKARAKA) and 81-96 (PRHLQLAIRNDEELNK) of human Histone H2A. | |
| 0.1 mg | |
| Chromatin Research, Epigenetics, Histones and Modified Histones | |
| 8338 | |
| Human, Mouse, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction