missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histone H2A.Z Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116156
This item is not returnable.
View return policy
Description
Histone H2A.Z Polyclonal Antibody for Western Blot, IHC (P)
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.
Specifications
| Histone H2A.Z | |
| Polyclonal | |
| Unconjugated | |
| H2AZ1 | |
| 11ac; C530002L11Rik; H2A histone family member V; H2A histone family member Z; H2A histone family member Z L homeolog; H2A histone family, member V; H2A histone family, member Va; H2A histone family, member Z; H2A.F/Z; H2A.z; H2A.Z variant histone 1; H2A.Z variant histone 2; H2A.Z variant histone 2a; H2A.Z1; H2a.z-1; H2A.Z-2; H2A.ZK5; H2A.ZK5ac; H2A.Zl; h2a.zl1; h2a.zl2; H2A/z; H2AF; H2AFV; h2afva; h2afz; h2afz.L; H2AFZac; H2AV; H2az; H2AZ histone; H2AZ1; H2AZ2; h2az2a; h2aza; H2AZK11ac; H2AZK7ac; histone (H2A.Z); histone H2A.F/Z; Histone H2A.V; Histone H2A.Z; histone H2A.Zl2; histone H2A.Z-like; histone variant H2A.F/Z; hypothetical protein; purine-rich binding element protein B; RCJMB04_2h11; Unknown (protein for MGC:133654); unnamed protein product; XELAEV_18005732mg; XH2AZ | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 3015, 94239 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P0C0S5, Q71UI9 | |
| H2AFZ, H2AZ1, H2az2 | |
| KLH conjugated synthetic peptide between 59-86 amino acids from the C-terminal region of human H2AFZ. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction