Learn More
Histone H3.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-81964-0.025ML
 View more versions of this product
Product Code. 18245544
  • 215.00€ / 25µL
Description
Histone H3.1 Polyclonal specifically detects Histone H3.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H3.1
Polyclonal
Unconjugated
phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
H3 histone family, member D, H3.1HIST1H3D, H3/d, H3FA, H3FB, H3FC, H3FDHIST1H3F, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3e, histone cluster 1, H3e, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l
Rabbit
17 kDa
0.025 mL
Epigenetics, Histones and Modified Histones
8353
Rat, Human, Mouse
IgG
Immunofluorescence, Western Blot, Immunocytochemistry
1.0 mg/mL
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200
P68431
HIST1H3E
The antiserum was produced against synthesized non-phosphopeptide derived from human Histone H3.1 around aa. 10 (R-K-S-T-G).
Affinity Purified
RUO
Primary
Histone H3.1 (specific to a.a. 10) detects endogenous levels of total Histone H3.1 protein. NB100-81964 is also cross-reactive with Histone H3.2 and H3.3 from human mouse and rat.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.